missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terazosin EP Impurity M (Prazosin EP Impurity B), Mikromol™
Description
Terazosin EP Impurity M (Prazosin EP Impurity B)
Specifications
Specifications
| Chemical Name or Material | 1,4-Bis(furan-2-ylcarbonyl)piperazine |
| CAS | 31350-27-3 |
| Analyte or Component Names | 1,4-Bis(furan-2-ylcarbonyl)piperazine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H14 N2 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Prazosin Hydrochloride Imp. B (EP), Prazosin Imp. B (EP), Terazosin Hydrochloride Dihydrate Imp. M (EP), Terazosin Imp. M (EP), 1,4-Bis(furan-2-ylcarbonyl)piperazine, Prazosin Hydrochloride Impurity B, Terazosin Hydrochloride Dihydrate Impurity M, Prazosin Impurity B, Terazosin Impurity M |
| InChI Formula | InChI=1S/C14H14N2O4/c17-13(11-3-1-9-19-11)15-5-7-16(8-6-15)14(18)12-4-2-10-20-12/h1-4,9-10H,5-8H2 |
| SMILES | O=C(N1CCN(CC1)C(=O)c2occc2)c3occc3 |
| IUPAC Name | [4-(furan-2-carbonyl)piperazin-1-yl]-(furan-2-yl)methanone |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?