missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TER119 Rat, anti-Mouse, PE, Clone: TER-119, STEMCELL Technologies
Rat Monoclonal Antibody
£101.25 - £255.15
Specifications
| Antigen | TER119 |
|---|---|
| Clone | TER-119 |
| Applications | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17187611
|
Stemcell Technologies
100-0451 |
25 μg |
£101.25
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17197611
|
Stemcell Technologies
100-0452 |
100 μg |
£255.15
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The TER-119 antibody reacts with murine TER119 (Ly-76), an ∼52 kDa protein associated with glycophorin A on the surface of cells of the erythroid lineage in embryonic yolk sac, fetal and newborn liver, adult bone marrow, peripheral blood, and lymphoid organs
- TER119 is an erythroid-specific marker expressed at all stages of differentiation from early proerythroblasts to mature erythrocytes, but not by erythroid colony-forming cells (BFU-E, blast-forming unit erythroid, or CFU-E, colony-forming unit erythroid)
- The TER-119 antibody is a component of the 'lineage cocktail' used to detect, or deplete cells committed to hematopoietic lineages
- In adult mice, TER119 is found on ∼20 - 25% of bone marrow cells and ∼2 - 3% of splenocytes
- This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse CD4+ T Cell Isolation Kit (Catalog #19852)
Specifications
| TER119 | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| PE | |
| Mouse |
| TER-119 | |
| Monoclonal | |
| Rat | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title