Learn More
Invitrogen™ TER-119 Monoclonal Antibody (TER-119), Brilliant Ultra Violet™ 805, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£176.00 - £353.00
Specifications
| Antigen | TER-119 |
|---|---|
| Clone | TER-119 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17473532
![]() |
Invitrogen™
368-5921-82 |
100 μg |
£353.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17463532
|
Invitrogen™
368-5921-80 |
25 μg |
£176.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TER-119 monoclonal antibody reacts with mouse erythroid cells from early proerythroblast to mature erythrocyte stages. The TER-119 antigen is present in yolk sac, fetal and newborn liver, but is not expressed by cells carrying BFU-E and CFU-E activities. Several erythroleukemia cell lines tested so far are negative for expression of TER-119 antigen even after dimethylsulfoxide stimulation. Biochemical and molecular analysis of the TER-119 antigen indicate that this molecule is associated with the surface glycophorin A, but is not a typical glycophorin. Applications Reported: This TER-119 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TER-119 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This may be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Brilliant Ultraviolet 805 is a tandem dye that emits at 797 nm and is intended for use on cytometers equipped with an ultraviolet (355 nm) laser.
TER-119 is a lineage marker for erythroid cells from early proerythroblast to mature erythrocyte stages in adult blood, spleen, and bone marrow. It is also present in yolk sac, and fetal and newborn liver. The TER-119 antigen is not expressed by cells of earlier erythroid development at BFU-e (blast-forming unit erythroid) stage or CFU-e (colony-forming unit erythroid) stage.Specifications
| TER-119 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Ly76; Ly-76; TER119 | |
| Ly76 | |
| Primary | |
| Antibody |
| TER-119 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 805 | |
| Rat | |
| Mouse | |
| 104231 | |
| IgG2b κ | |
| Affinity chromatography | |
| Ly76 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.