missing translation for 'onlineSavingsMsg'
Learn More

TEPP, TRC

Product Code. 30422230
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422230 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422230 Supplier Toronto Research Chemicals Supplier No. TRCT10010025MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O,O-TEPP
CAS 107-49-3
Molecular Formula C8 H20 O7 P2
Quantity 25 mg
Synonym Diphosphoric acid','P,P,P',P'-tetraethyl ester, Diphosphoric acid','tetraethyl ester (9CI), Pyrophosphoric acid','tetraethyl ester (8CI), Bis-O,O-diethylphosphoric anhydride, Bladan, Ethyl pyrophosphate (Et4P2O7), Killex, NSC 14707, NSC 646213, Nifos, Pyrodust, TEP, TEP (phosphate), TEPP, Tetraethyl diphosphate, Tetraethyl pyrophosphate, Tetrastigmine, Vapotone, O,O-TEPP
InChI Formula InChI=1S/C8H20O7P2/c1-5-11-16(9,12-6-2)15-17(10,13-7-3)14-8-4/h5-8H2,1-4H3
SMILES CCOP(=O)(OCC)OP(=O)(OCC)OCC
IUPAC Name diethoxyphosphoryl diethyl phosphate
Molecular Weight (g/mol) 290.19
Formula Weight 290.0684
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.