missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEPP, TRC
Specifications
Specifications
| Chemical Name or Material | O,O-TEPP |
| CAS | 107-49-3 |
| Molecular Formula | C8 H20 O7 P2 |
| Quantity | 100 mg |
| Synonym | Diphosphoric acid','P,P,P',P'-tetraethyl ester, Diphosphoric acid','tetraethyl ester (9CI), Pyrophosphoric acid','tetraethyl ester (8CI), Bis-O,O-diethylphosphoric anhydride, Bladan, Ethyl pyrophosphate (Et4P2O7), Killex, NSC 14707, NSC 646213, Nifos, Pyrodust, TEP, TEP (phosphate), TEPP, Tetraethyl diphosphate, Tetraethyl pyrophosphate, Tetrastigmine, Vapotone, O,O-TEPP |
| InChI Formula | InChI=1S/C8H20O7P2/c1-5-11-16(9,12-6-2)15-17(10,13-7-3)14-8-4/h5-8H2,1-4H3 |
| SMILES | CCOP(=O)(OCC)OP(=O)(OCC)OCC |
| IUPAC Name | diethoxyphosphoryl diethyl phosphate |
| Molecular Weight (g/mol) | 290.19 |
| Formula Weight | 290.0684 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?