missing translation for 'onlineSavingsMsg'
Learn More

TEPP-46, MedChemExpress

Product Code. 16493577
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493577 50 mg 50mg
16487558 10 mg 10mg
16487496 100 mg 100mg
16450038 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16493577 Supplier Medchem Express Supplier No. HY18657

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TEPP-46 (ML-265) is a potent and selective pyruvate kinase M2 (PKM2) activator with an AC50 of 92 nM, showing little or no effect on PKM1, PKL and PKR.

TEPP-46 (ML-265) is a potent and selective pyruvate kinase M2 (PKM2) activator with an AC50 of 92 nM, showing little or no effect on PKM1, PKL and PKR.

Specifications

Chemical Name or Material TEPP-46
CAS 1221186-53-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C17H16N4O2S2
Quantity 50 mg
Synonym ML-265
Solubility Information DMSO : 50 mg/mL (134.24 mM; Need ultrasonic)
SMILES O=C1C(N(C)C2=C3SC(S(C)=O)=C2)=C3C=NN1CC4=CC=CC(N)=C4
Molecular Weight (g/mol) 372.46
Formula Weight 372.46
Percent Purity 95.86%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.