missing translation for 'onlineSavingsMsg'
Learn More

TEP1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18410764 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410764 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410764 Supplier Novus Biologicals Supplier No. NBP177285AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TEP1 Polyclonal antibody specifically detects TEP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TEP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias p240telomerase protein component 1, telomerase-associated protein 1VAULT2, TLP1p80 telomerase homolog, TP1Telomerase protein 1, TROVE domain family, member 1, TROVE1
Host Species Rabbit
Immunogen Antibody was raised against a 20 amino acid synthetic peptide from near the amino terminus of human TP1. The immunogen is located within the first 50 amino acids of TP1. Amino Acid Squence: EKLHGHVSAHPDILSLENRC
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 7011
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.