missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TentaGel™ S-NH2, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 354610050
118.83 GBP valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For solid support synthesisSpecifications
TentaGel™ S-NH2 | |
Powder | |
Glass bottle | |
5 g | |
O-(2-Aminoethyl)polyethylene glycol polymer bound |
White to Yellow | |
0.2 to 0.3mmol/g | |
90 Micron | |
MFCD00217899 |
RUO – Research Use Only