missing translation for 'onlineSavingsMsg'
Learn More

TentaGel™ MB-S-Trityl, Thermo Scientific™

Brand:  Thermo Scientific Acros 354550010

Code : 29

Additional Details : Weight : 0.00100kg

 View more versions of this product

Product Code. 10633562

  • £27.60 / 1g
Estimated Shipment: 16-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

For solid support synthesis
Specifications

Specifications

White to Yellow
Powder
Glass bottle
1 g
O-[2-(3-Tritylthiopropionamido)ethyl]polyethylene glycol polymer bound
TentaGel™ MB-S-Trityl
0.4 to 0.6mmol/g
140 to 170 Micron
MFCD01868636
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only