missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tensin 4 Recombinant Rabbit Monoclonal Antibody (23GB4225), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£162.00 - £387.00

Specifications

Antigen Tensin 4
Clone 23GB4225
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30088039
GREENER_CHOICE
View Documents
Invitrogen™
MA552666
100 μL
£387.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30089677
View Documents
Invitrogen™
MA552667
20 μL
£162.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tensin 4 Recombinant Monoclonal antibody specifically detects Tensin 4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications

Specifications

Tensin 4
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse, Rat
Q4V8I3, Q8BZ33, Q8IZW8
217169, 303517, 84951
IgG
Affinity chromatography
Antibody
23GB4225
Recombinant Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
9930017A07Rik; AA589547; AU016405; C terminal tensin like; CTEN; c-ten; C-terminal tensin-like protein; PP14434; RGD1310402; tensin 4; tensin-4; TNS4
A synthesized peptide derived from TNS4 (200-233AA).
Primary
-20°C
TNS4
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.