missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tensin 4 Monoclonal Antibody (684524)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524311
This item is not returnable.
View return policy
Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
TNS4 links the focal adhesion and cell adhesion signaling to the actin cytoskeleton. It may be involved in cell survival and migration. The protein may be involved in cell migration, cartilage development and in linking signal transduction pathways to the cytoskeleton and promote apoptosis, via its cleavage by caspase-3.
Specifications
| Tensin 4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8IZW8 | |
| TNS4 | |
| E. coli-derived recombinant human Tensin 4/CTEN Ser2-Gln107. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 684524 | |
| Unconjugated | |
| TNS4 | |
| 9930017A07Rik; AA589547; AU016405; C terminal tensin like; CTEN; c-ten; C-terminal tensin-like protein; PP14434; RGD1310402; tensin 4; tensin-4; TNS4 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 84951 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction