missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tensin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557516
This item is not returnable.
View return policy
Description
Immunogen sequence: EEPLNLEGLV AHRVAGVQAR EKQPAEPPAP LRRRAASDGQ YENQSPEATS PRSPGVRSPV QCVSPELALT IALNPGGRPK EPHLHSYK Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.
Tensin is involved in the maintenance of cellular structure by anchoring Actin filaments at the focal adhesion via F-Actin binding and capping activities. However, tensin also contains a Src homology 2 (SH2) domain and has the ability to be phosphorylated. Tensin is phosphorylated on tyrosine, serine, and threonine residues, suggesting that it might participate in signal transduction cascades. These diverse characteristics in a single molecule indicate that tensin may be an important link between the cytoskeleton and signal transduction pathways.
Specifications
| Tensin 1 | |
| Polyclonal | |
| Unconjugated | |
| TNS1 | |
| 1110018I21Rik; 1200014E20Rik; AI648117; DKFZp586K0617; E030018G17Rik; E030037J05Rik; matrix-remodelling associated 6; Matrix-remodelling-associated protein 6; MGC88584; MST091; MST122; MST127; MSTP091; MSTP122; MSTP127; MXRA6; PPP1R155; protein phosphatase 1, regulatory subunit 155; tensin 1; tensin-1; TNS; TNS1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7145 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HBL0 | |
| TNS1 | |
| Recombinant protein corresponding to Human Tensin 1. Recombinant protein control fragment (Product #RP-96507). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction