missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tensin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567975
This item is not returnable.
View return policy
Description
Predicted to react with Mouse samples.
Tensin is involved in the maintenance of cellular structure by anchoring Actin filaments at the focal adhesion via F-Actin binding and capping activities. However, tensin also contains a Src homology 2 (SH2) domain and has the ability to be phosphorylated. Tensin is phosphorylated on tyrosine, serine, and threonine residues, suggesting that it might participate in signal transduction cascades. These diverse characteristics in a single molecule indicate that tensin may be an important link between the cytoskeleton and signal transduction pathways.
Specifications
| Tensin 1 | |
| Polyclonal | |
| Unconjugated | |
| TNS1 | |
| 1110018I21Rik; 1200014E20Rik; AI648117; DKFZp586K0617; E030018G17Rik; E030037J05Rik; matrix-remodelling associated 6; Matrix-remodelling-associated protein 6; MGC88584; MST091; MST122; MST127; MSTP091; MSTP122; MSTP127; MXRA6; PPP1R155; protein phosphatase 1, regulatory subunit 155; tensin 1; tensin-1; TNS; TNS1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 21961, 7145 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9HBL0 | |
| TNS1 | |
| Synthesized peptide derived from human Tensin 1. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction