missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tensin 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£305.00 - £451.00
Specifications
| Antigen | Tensin 1 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18475940
|
Novus Biologicals
NBP1-84130-25ul |
25 μL |
£305.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18710734
|
Novus Biologicals
NBP1-84130 |
£451.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Tensin 1 Polyclonal specifically detects Tensin 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| Tensin 1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| RUO | |
| Human | |
| DKFZp586K0617, matrix-remodelling associated 6, Matrix-remodelling-associated protein 6, MGC88584, MST091, MST122, MST127, MSTP127, MXRA6, tensin, tensin 1, tensin-1, TNSMSTP122 | |
| TNS1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication, Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 7145 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LSPLTNGLDKSYPMEPMVNGGGYPYESASRAGPAHAGHTAPMRPSYSAQEGLAGYQREGPHPAWPQPVTTSHYAHDPSGMFRSQS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title