missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TENR Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £364.00
Specifications
| Antigen | TENR |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TENR Polyclonal antibody specifically detects TENR in Human, Rat samples. It is validated for Western BlotSpecifications
| TENR | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Stem Cells | |
| PBS (pH 7.3), 50% glycerol | |
| 132612 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| adenosine deaminase domain containing 1 (testis-specific), adenosine deaminase domain-containing protein 1, FLJ32741, TENR, Testis nuclear RNA-binding protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human ADAD1 (NP_001152757.1). MASNNHWFQSSQVPSFAQMLKKNLPVQPATKTITTPTGWSSESYGLSKMASKVTQVTGNFPEPLLSKNLSSISNPVLPPKKIPKEFIMKYKRGEINPVSALHQFAQMQRVQLDLKETVTTGNVMGPYFAFCAVVDGIQYKTGLGQNKKESRSNAAKLALDELLQLDEPEPRILETSGPPPFPAEPVVLSELAYVSKVHYEGRHIQYAKISQIVKERFNQLISNRSEYLKYSSSLAAFIIERAGQHEVVAIGTGEYNYSQDIKPDGRVLHDTHAVVTARRS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title