missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pirbuterol-(tert-butyl-d9) acetate, Honeywell Fluka™
Click to view available options
Quantity:
10 mg
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
Specifications
CAS | 1431291-46-1 |
Packaging | Glass Bottle |
Quantity | 10 mg |
Grade | Analytical Standard |
For Use With (Application) | Beta-agonists and Antagonists |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'