missing translation for 'onlineSavingsMsg'
Learn More

Tenovin-1, MedChemExpress

Product Code. 16426186
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16426186

Brand: Medchem Express HY13423

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tenovin-1, a p53 activator, protects p53 from MDM2-mediated degradation. Tenovin-1 acts through inhibition of the protein-deacetylating activities of SirT1 and SirT2. Tenovin-1 is also a dihydroorotate dehydrogenase (DHODH) inhibitor.

Tenovin-1, a p53 activator, protects p53 from MDM2-mediated degradation. Tenovin-1 acts through inhibition of the protein-deacetylating activities of SirT1 and SirT2. Tenovin-1 is also a dihydroorotate dehydrogenase (DHODH) inhibitor.

Specifications

Chemical Name or Material Tenovin-1
Physical Form Solid
CAS 380315-80-0
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H23N3O2S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 100 mg/mL (270.65 mM; Need ultrasonic)
SMILES O=C(NC(NC1=CC=C(NC(C)=O)C=C1)=S)C2=CC=C(C(C)(C)C)C=C2
Molecular Weight (g/mol) 369.48
Formula Weight 369.48
Percent Purity 99.79%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.