missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenovin-1, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Description
Tenovin-1, a p53 activator, protects p53 from MDM2-mediated degradation. Tenovin-1 acts through inhibition of the protein-deacetylating activities of SirT1 and SirT2. Tenovin-1 is also a dihydroorotate dehydrogenase (DHODH) inhibitor.
Specifications
Specifications
| Chemical Name or Material | Tenovin-1 |
| Physical Form | Solid |
| CAS | 380315-80-0 |
| Color | Off-White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C20H23N3O2S |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : 100 mg/mL (270.65 mM; Need ultrasonic) |
| SMILES | O=C(NC(NC1=CC=C(NC(C)=O)C=C1)=S)C2=CC=C(C(C)(C)C)C=C2 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction