missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenofovir, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Tenofovir
Specifications
Specifications
| Chemical Name or Material | Tenofovir |
| CAS | 147127-20-6 |
| Analyte or Component Names | Tenofovir |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C9 H14 N5 O4 P |
| Certifications/Compliance | ISO 9001 |
| Synonym | [[(1R)-2(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]phosphonic Acid |
| InChI Formula | InChI=1S/C9H14N5O4P/c1-6(18-5-19(15,16)17)2-14-4-13-7-8(10)11-3-12-9(7)14/h3-4,6H,2,5H2,1H3,(H2,10,11,12)(H2,15,16,17)/t6-/m1/s1 |
| SMILES | C[C@H](Cn1cnc2c(N)ncnc12)OCP(=O)(O)O |
| IUPAC Name | [(2R)-1-(6-aminopurin-9-yl)propan-2-yl]oxymethylphosphonic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction