missing translation for 'onlineSavingsMsg'
Learn More

Tenofovir, MedChemExpress

Product Code. 16446356
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16446356

Brand: Medchem Express HY13910

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tenofovir (GS 1278) is a nucleotide reverse transcriptase inhibitor to treat HIV and chronic Hepatitis B (HBV).

Tenofovir (GS 1278) is a nucleotide reverse transcriptase inhibitor to treat HIV and chronic Hepatitis B (HBV).

Specifications

Chemical Name or Material Tenofovir
CAS 147127-20-6
Color White
Purity Grade Notes Research
Molecular Formula C9H14N5O4P
Quantity 100 mg
Synonym GS 1278 PMPA
Solubility Information H2O : 2 mg/mL (6.96 mM; Need ultrasonic)
SMILES C[C@@H](OCP(O)(O)=O)CN1C=NC2=C(N)N=CN=C12
Molecular Weight (g/mol) 287.21
Formula Weight 287.21
Percent Purity 98.4%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.