missing translation for 'onlineSavingsMsg'
Learn More

Tenofovir hydrate, MedChemExpress

Product Code. 16428547
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16428547 5 mg 5mg
16455908 10 mg 10mg
16442057 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16428547 Supplier Medchem Express Supplier No. HY13910A5mg
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tenofovir hydrate is a nucleotide reverse transcriptase inhibitor to treat HIV and chronic Hepatitis B.

Tenofovir hydrate is a nucleotide reverse transcriptase inhibitor to treat HIV and chronic Hepatitis B.

Specifications

Chemical Name or Material Tenofovir hydrate
CAS 206184-49-8
Color White
Purity Grade Notes Research
Molecular Formula C9H16N5O5P
Quantity 5 mg
Synonym GS 1278 hydratePMPA hydrate
Solubility Information DMSO : ≥ 6 mg/mL (19.66 mM)
SMILES C[C@@H](OCP(O)(O)=O)CN1C=NC2=C(N)N=CN=C12.O
Molecular Weight (g/mol) 305.23
Formula Weight 305.23
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.