missing translation for 'onlineSavingsMsg'
Learn More

Tenofovir disoproxil fumarate, 98%, Thermo Scientific Chemicals

Product Code. 15668725 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15668725 1 g 1g
15678725 5 g 5g
15688725 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15668725 Supplier Thermo Scientific Acros Supplier No. 461250010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202138-50-9
Molecular Formula C19H30N5O10P·C4H4O4
Molecular Weight (g/mol) 635.51
InChI Key VCMJCVGFSROFHV-WZGZYPNHSA-N
Synonym tenofovir disoproxil fumarate, viread, tenofovir df, unii-ott9j7900i, tenofovir disoproxil fumarate usan, 9-r-2-bis isopropoxycarbonyl oxy methoxy phosphinyl methoxy propyl adenine fumarate, 9-r-2-bis isopropoxycarbonyl oxy methoxy phosphinyl methoxy propyl adenine, fumarate, r-5-2-6-amino-9h-purin-9-yl-1-methylethoxy methyl-2,4,6,8-tetraoxa-5-phosphanonanedioic acid, bis 1-methylethyl ester, 5-oxide, e-2-butenedioate 1:1, 9-r-2-bis isopropoxycarbonyl oxy methoxy phosphinyl methoxy propyl adenine fumarate 1:1
PubChem CID 6398764
ChEBI CHEBI:63718
IUPAC Name [[(2R)-1-(6-aminopurin-9-yl)propan-2-yl]oxymethyl-(propan-2-yloxycarbonyloxymethoxy)phosphoryl]oxymethyl propan-2-yl carbonate;(E)-but-2-enedioic acid
SMILES CC(C)OC(=O)OCOP(=O)(COC(C)CN1C=NC2=C1N=CN=C2N)OCOC(=O)OC(C)C.C(=CC(=O)O)C(=O)O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: >10 mg/mL.
Formula Weight 635.51
Percent Purity 98%
Chemical Name or Material Tenofovir disoproxil fumarate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.