missing translation for 'onlineSavingsMsg'
Learn More

Tenofovir Disoproxil Dimer, TRC

Product Code. 30011113
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011113 50 mg 50mg
30004418 25 mg 25mg
17981265 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011113 Supplier Toronto Research Chemicals Supplier No. TRCT01851550MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1093279-76-5
Molecular Formula C39 H60 N10 O20 P2
Molecular Weight (g/mol) 1050.9
Synonym 2,4,6,8-Tetraoxa-5-phosphanonanedioic acid, 5-[[(1R)-2-[6-[[[[9-[(2R)-2,11-dimethyl-5-[[[(1-methylethoxy)carbonyl]oxy]methoxy]-5-oxido-9-oxo-3,6,8,10-tetraoxa-5-phosphadodec-1-yl]-9H-purin-6-yl]amino]methyl]amino]-9H-purin-9-yl]-1-methylethoxy]methyl]-, 1,9-bis(1-methylethyl) ester, 5-oxide, Tenofovir Disoproxil Dimer
IUPAC Name [[(2R)-1-[6-[[[9-[(2R)-2-[bis(propan-2-yloxycarbonyloxymethoxy)phosphorylmethoxy]propyl]purin-6-yl]amino]methylamino]purin-9-yl]propan-2-yl]oxymethyl-(propan-2-yloxycarbonyloxymethoxy)phosphoryl]oxymethyl propan-2-yl carbonate
SMILES CC(C)OC(=O)OCOP(=O)(CO[C@H](C)Cn1cnc2c(NCNc3ncnc4c3ncn4C[C@@H](C)OCP(=O)(OCOC(=O)OC(C)C)OCOC(=O)OC(C)C)ncnc12)OCOC(=O)OC(C)C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C39H60N10O20P2/c1-24(2)66-36(50)56-18-62-70(54,63-19-57-37(51)67-25(3)4)22-60-28(9)11-48-16-46-30-32(42-14-44-34(30)48)40-13-41-33-31-35(45-15-43-33)49(17-47-31)12-29(10)61-23-71(55,64-20-58-38(52)68-26(5)6)65-21-59-39(53)69-27(7)8/h14-17,24-29H,11-13,18-23H2,1-10H3,(H,40,42,44)(H,41,43,45)/t28-,29-/m1/s1
Formula Weight 1050.3461
Chemical Name or Material Tenofovir Disoproxil Dimer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.