Learn More
Invitrogen™ TENM1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590694
Description
Immunogen sequence: QPVEGELYAN GVSKGNRGTE SMDTTYSPIG GKVSDKSEKK VFQKGRAIDT GEVDIGAQVM QTIPPGLFWR FQITIHHPIY LKFNISLAKD SLLGIYGRRN IPPTHTQFDF VKLMDGKQLV KQDSKGSDDT QHSPRNLILT SLQETGFIEY MDQGPWYLAF YNDGKKMEQV FVLTTAIEIM DDCSTNCNGN G; Positive Samples: U-87MG, U-251MG, HeLa; Cellular Location: Cell membrane, Cytoplasm, Nucleus, Nucleus matrix, Nucleus speckle, Single-pass membrane protein, cytoskeleton.
The teneurin transmembrane protein 1 (TENM1) is a member of a family of four neuronal cell surface proteins homologous to the Drosophila pair-rule gene Ten-m. TENM1 is expressed primarily in the developing central nervous system and may be proteolytically cleaved with the intracellular domain translocating to the nucleus. TENM1 is a direct target of the homeobox transcription factor EMX2, a transcription factor thought to be important for area specification in the developing cortex.
Specifications
| TENM1 | |
| Polyclonal | |
| Unconjugated | |
| TENM1 | |
| IDten-1; odz (odd Oz/ten-m, Drosophila) homolog 3; odz, odd Oz/ten-m homolog 1; odz, odd Oz/ten-m homolog 1(Drosophila); Odz1; Odz2; ODZ3; Protein Odd Oz/ten-m homolog 1; RGD1561678; TCAP-1; TCPA-1; Ten-1; Ten-1 ECD; Ten-1 extracellular domain; Ten-1 intracellular domain; tenascin M1; tenascin-M1; teneurin 1; Teneurin C-terminal-associated peptide; Teneurin transmembrane protein 1; teneurin-1; TENM1; Ten-m1; TNM; TNM1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10178, 23963, 298168 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.25 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9UKZ4, Q9WTS4 | |
| TENM1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 350-540 of human TENM1 (NP_0011567501). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.