missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TENM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551753
This item is not returnable.
View return policy
Description
Immunogen sequence: IRTISRNQAH LNDMNIYEIA SPADQELYQF TVNGTHLHTL NLITRDYVYN FTYNSEGDLG AITSSNGNSV HIRRDAGGMP LWLVVPGGQV YWLTISSNGV LKRVSAQGYN LALMTYPGNT GLLATKSNEN GWTTVYEYDP EGH Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 61%.
The teneurin transmembrane protein 1 (TENM1) is a member of a family of four neuronal cell surface proteins homologous to the Drosophila pair-rule gene Ten-m. TENM1 is expressed primarily in the developing central nervous system and may be proteolytically cleaved with the intracellular domain translocating to the nucleus. TENM1 is a direct target of the homeobox transcription factor EMX2, a transcription factor thought to be important for area specification in the developing cortex.
Specifications
| TENM1 | |
| Polyclonal | |
| Unconjugated | |
| TENM1 | |
| IDten-1; odz (odd Oz/ten-m, Drosophila) homolog 3; odz, odd Oz/ten-m homolog 1; odz, odd Oz/ten-m homolog 1(Drosophila); Odz1; Odz2; ODZ3; Protein Odd Oz/ten-m homolog 1; RGD1561678; TCAP-1; TCPA-1; Ten-1; Ten-1 ECD; Ten-1 extracellular domain; Ten-1 intracellular domain; tenascin M1; tenascin-M1; teneurin 1; Teneurin C-terminal-associated peptide; Teneurin transmembrane protein 1; teneurin-1; TENM1; Ten-m1; TNM; TNM1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10178 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UKZ4 | |
| TENM1 | |
| Recombinant protein corresponding to Human TENM1. Recombinant protein control fragment (Product #RP-92160). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction