missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Teniposide

Product Code. 18766236
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18766236 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18766236 Supplier R&D Systems™ Supplier No. 6975/100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. DNA topoisomerase II inhibitor; viral transduction enhancer

Teniposide is a DNA topoisomerase II inhibitor and viral transduction enhancer. It induces breaks in double-stranded DNA and prevents DNA repair. Teniposide enhances transduction of recombinant adeno-associated virus 2 (rAAV2) by up to 40-fold in human cells in vitro and by 100-fold in vivo at 24 h post-transduction.
TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material (5 R,5 aR,8 aR,9 S)-5,8,8 A,9-Tetrahydro-5-(4-hydroxy-3,5-dimethoxyphenyl)-9-[[4,6-O-[(R)-2-thienylmethylene]-β-D-glucopyranosyl]oxy]furo[3 ',4 ':6,7 ]naphtho[2,3-d]-1,3-dioxol-6(5 aH)-one
CAS 29767-20-2
Quantity 100 mg
Product Line Tocris
Target DNA Topoisomerase Inhibitors
Molecular Formula C32H32O13S
Purity 0.98
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.