missing translation for 'onlineSavingsMsg'
Learn More

Teneligliptin hydrobromide, MedChemExpress

Product Code. 16446556
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
50mg
This item is not returnable. View return policy

Product Code. 16446556

Brand: Medchem Express HY14806A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Teneligliptin (MP-513) hydrobromide is a potent chemotype prolylthiazolidine-based DPP-4 inhibitor, which competitively inhibits human plasma, rat plasma, and human recombinant DPP-4 in vitro, with IC50s of approximately 1 nM.

Teneligliptin (MP-513) hydrobromide is a potent chemotype prolylthiazolidine-based DPP-4 inhibitor, which competitively inhibits human plasma, rat plasma, and human recombinant DPP-4 in vitro, with IC50s of approximately 1 nM.

Specifications

Chemical Name or Material Teneligliptin hydrobromide
CAS 906093-29-6
Color Colorless
Purity Grade Notes Research
Molecular Formula C22H30N6OS.5/2HBr
Quantity 100 mg
Synonym MP-513 hydrobromide
Solubility Information H2O : ≥ 200 mg/mL (318.04 mM) ∣DMSO : ≥ 100 mg/mL (159.02 mM)
SMILES O=C(N1CSCC1)[C@H]2NC[C@@H](N3CCN(C4=CC(C)=NN4C5=CC=CC=C5)CC3)C2.[2.5 HBr]
Molecular Weight (g/mol) 628.86
Formula Weight 628.86
Percent Purity 98.71%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.