missing translation for 'onlineSavingsMsg'
Learn More

Tenax™ TA, 80-100 mesh

Product Code. 16554354 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16554354 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16554354 Supplier Thermo Scientific Acros Supplier No. 462210100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24938-68-9 | C20H18O | 274.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 24938-68-9
Melting Point 460.0°C
Molecular Formula C20H18O
Quantity 10 g
InChI Key TWADJGWUKGOPFG-UHFFFAOYSA-N
SMILES COC1=C(C=C(C)C=C1C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name 2-methoxy-5-methyl-3-phenyl-1,1'-biphenyl
Molecular Weight (g/mol) 274.36
Chemical Name or Material Tenax* TA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.