missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenascin-X Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13595-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is raised against human TNXB and detects a band about 75 kDa, which probably represent a proteolytic cleaved form of human TNXB.
Tenascin-X (TNXB) is a extracellular matrix glycoprotein predomitly located in the outer reticular lamina of the basement membrane (BM). It interacts with many other ECM proteins and it accelerates collagen fibrillogenesisin vitro. TNXB plays a role in interactions between cell and ECM, antiadhesive effect, inhibiting cell migration, and in maintaining homeostasis of the extracellular matrix. Deficiency of TNXB has been associated with the connective tissue disorder Ehlers-Danlos syndrome.Specifications
| Tenascin-X | |
| Polyclonal | |
| Unconjugated | |
| TNXB | |
| Hexabrachion like protein, HXBL, Tenascin X, tenascin XB, TENX, TN X, TNX, TNXB, TNXB1, TNXB2, TNXBS, XB, XBS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 415089, 7148, 81877 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P22105 | |
| Tnxb | |
| Tenascin-X Fusion Protein Ag4501 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction