missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenascin X Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£199.00 - £255.00
Specifications
| Antigen | Tenascin X |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18674725
|
Novus Biologicals
NBP2-99638-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18655455
|
Novus Biologicals
NBP2-99638-50ul |
50 μg |
£199.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tenascin X Polyclonal antibody specifically detects Tenascin X in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Tenascin X | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cellular Markers, Extracellular Matrix | |
| 0.2 um filtered solution in PBS | |
| 7148 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Hexabrachion-like protein, HXBL, tenascin XB, tenascin XB2, tenascin-X, TNXB1growth-inhibiting protein 45, TNXB2tenascin XB1, TNXBSTNXTENX, XBS, XBTN-X | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Tenascin X. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title