missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenascin X Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21427DL594
This item is not returnable.
View return policy
Description
Tenascin X Polyclonal antibody specifically detects Tenascin X in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Tenascin X | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Hexabrachion-like protein, HXBL, tenascin XB, tenascin XB2, tenascin-X, TNXB1growth-inhibiting protein 45, TNXB2tenascin XB1, TNXBSTNXTENX, XBS, XBTN-X | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Tenascin X | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Extracellular Matrix | |
| 7148 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction