missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tenascin R Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550580
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total Tenascin R protein.
Tenascin-R (TNR) is an extracellular matix protein expressed primarily in the central nervous system. It is a member of the tenascin (TN) gene family, which includes at least 3 genes in mammals: TNC (or hexabrachion; MIM 187380), TNX (TNXB; MIM 600985), and TNR (Erickson, 1993). The genes are expressed in distinct tissues at different times during embryonic development and are present in adult tissues.
Specifications
| Tenascin R | |
| Polyclonal | |
| Unconjugated | |
| Tnr | |
| J1NRM; J1-tenascin; janusin; Neural recognition molecule J1-160/180; restrictin; tenascin R; tenascin R (restrictin, janusin); tenascin-R; Tenascin-R (Restrictin janusin J1-160/180); Tenascin-R (Restrictin, janusin, J1-160/180); Tnr; TN-R | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 21960, 25567, 7143 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 2.2 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q05546, Q8BYI9, Q92752 | |
| Tnr | |
| Synthetic peptide corresponding to a region derived from internal residues of human tenascin R. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction