missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tenascin C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119455
This item is not returnable.
View return policy
Description
Tenascin C Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Tenascin is a high molecular weight, multifunctional, extracellular matrix glycoprotein expressed in association with mesenchymal-epithelial interactions during development and in the neovasculature and stroma of undifferentiated tumors (Erickson and Bourdon 1989). It has been described under a variety of names: cytotactin, hexabrachion protein, J1, myotendinous antigen (MI) and glioma mesenchymal extracellular matrix (GMEM). The tenascin molecule is a disulfide-linked hexamer. The expression of tenascin is associated with development and growth, both normal and pathological, whereas the distribution in normal adult tissue is restricted.
Specifications
| Tenascin C | |
| Polyclonal | |
| Unconjugated | |
| TNC | |
| 150-225; AI528729; C130033P17Rik; cytotactin; deafness, autosomal dominant 56; DFNA56; Glioma-associated-extracellular matrix antigen; GMEM; GP; GP 150-225; hexabrachion; hexabrachion (tenascin); Hxb; JI; MGC144208; MGC144209; MGC167029; Myotendinous antigen; neuronectin; Ten; Tenascin; tenascin C; Tenascin-C; tenascin-C additional domain 1; tenascin-C isoform 14/AD1/16; TN; tnc; TN-C | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 116640 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| 0 | |
| TNC | |
| Recombinant protein Tenascin C. | |
| 200 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction