missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenascin C Antibody (4C8MS), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-68136AF700
This item is not returnable.
View return policy
Description
Tenascin C Monoclonal specifically detects Tenascin C in Human, Mouse, Rat, Feline samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Tenascin C | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| 150-225, Cytotactin, Glioma-associated-extracellular matrix antigen, GMEM, GP 150-225, Hexabrachion, hexabrachion (tenascin C, cytotactin), hexabrachion (tenascin), HXBcytotactin, JI, MGC167029, Myotendinous antigen, neuronectin, tenascin, tenascin C, Tenascin-C, tenascin-C isoform 14/AD1/16, TN-C, TNGP | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| NB110-68136 specifically reacts with Domain B on FNIII repeats of Tenascin C. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 4C8MS | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| P24821 | |
| TNC | |
| Recombinant human Tenascin C [Swiss-Prot# P24821]. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Extracellular Matrix, Neuroscience, Prostate Cancer | |
| 3371 | |
| Protein A purified | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction