missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Tenacissoside H

Product Code. 16479058
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479058 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16479058 Supplier MedChemExpress Supplier No. HYN0670
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Tenacissoside H is a Chinese medicine monomer extracted, isolated from Caulis Marsdeniae Tenacissimae.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
Molecular Weight (g/mol) 794.97 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 191729-45-0
Product Type Natural Products
Solubility DMSO : 100 mg/mL (125.79 mM∣Need ultrasonic)
Synonym Tenacissimoside C
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.