missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Temsirolimus, 99+%

A Rapamycin derivative with anti-cancer activity that inhibits mTOR

Brand:  Thermo Scientific™ J63654.MC

Quantity 100 mg

 View more versions of this product

Product Code. 15445829

  • ONLINE EXCLUSIVE PRICE
    £560.00
    £504.65 / 100mg
    Save £55.35
    9% Off
 Restricted Item
Estimated Shipment: 24-08-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

162635-04-3
-30°C to -10°C
A Rapamycin derivative with anti-cancer activity that inhibits mTOR
≥99%
1030.29g/mol
Powder
White to Off-white

Specifications

100 mg
-30°C to -10°C
A Rapamycin derivative with anti-cancer activity that inhibits mTOR
C56H87NO16
≥99%
White to Off-white
Powder
162635-04-3
1030.29g/mol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.