missing translation for 'onlineSavingsMsg'
Learn More

TEMPO, TRC

Product Code. 30135868
Change view
Click to view available options
Quantity:
1 g
5 g
500 g
Unit Size:
1g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135868 500 g 500g
30133267 5 g 5g
30006302 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135868 Supplier Toronto Research Chemicals Supplier No. TRCT017850500G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2564-83-2
Molecular Formula C9 H18 N O
Molecular Weight (g/mol) 156.25
Synonym 1-Piperidinyloxy, 2,2,6,6-tetramethyl- (9CI, ACI), 2,2,6,6-Tetramethyl-1-piperidinyloxy (ACI), Piperidinooxy, 2,2,6,6-tetramethyl- (7CI, 8CI), 1,1,5,5-Tetramethylpentamethylene nitroxide, 1-Oxyl-2,2,6,6-tetramethylpiperidine, 2,2,6,6-Tetramethyl-1-oxylpiperidine, 2,2,6,6-Tetramethyl-1-piperadoxyl, 2,2,6,6-Tetramethyl-1-piperidine-N-oxyl, 2,2,6,6-Tetramethyl-1-piperidinoxyl, 2,2,6,6-Tetramethyl-1-piperidyloxy, 2,2,6,6-Tetramethylpiperidin-1-oxy, 2,2,6,6-Tetramethylpiperidin-1-oxyl radical, 2,2,6,6-Tetramethylpiperidin-N-oxyl, 2,2,6,6-Tetramethylpiperidine N-oxide, 2,2,6,6-Tetramethylpiperidine N-oxide radical, 2,2,6,6-Tetramethylpiperidine N-oxy, 2,2,6,6-Tetramethylpiperidine N-oxyl, 2,2,6,6-Tetramethylpiperidine N-oxyl radical, 2,2,6,6-Tetramethylpiperidine nitroxide, 2,2,6,6-Tetramethylpiperidine nitroxide radical, 2,2,6,6-Tetramethylpiperidine oxide, 2,2,6,6-Tetramethylpiperidine-1-oxyl, 2,2,6,6-Tetramethylpiperidino-1-oxy, 2,2,6,6-Tetramethylpiperidinooxy, 2,2,6,6-Tetramethylpiperidinooxy radical, 2,2,6,6-Tetramethylpiperidinooxyl, 2,2,6,6-Tetramethylpiperidinoxy, 2,2,6,6-Tetramethylpiperidinoxyl, 2,2,6,6-Tetramethylpiperidinoxyl radical, 2,2,6,6-Tetramethylpiperidinyl 1-oxide, 2,2,6,6-Tetramethylpiperidinyl-1-oxyl, 2,2,6,6-Tetramethylpiperidinyl-N-oxy, 2,2,6,6-Tetramethylpiperidinyloxy, 2,2,6,6-Tetramethylpiperidoxyl, HO 6, T 3751, Tanan, Tanane, Tempo, TMPO
IUPAC Name 1-λ^{1}-oxidanyl-2,2,6,6-tetramethylpiperidine
SMILES CC1(C)CCCC(C)(C)N1[O]

Specifications

Quantity 500 g
InChI Formula InChI=1S/C9H18NO/c1-8(2)6-5-7-9(3,4)10(8)11/h5-7H2,1-4H3
Formula Weight 156.1388
Percent Purity >95%
Chemical Name or Material TEMPO
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.