missing translation for 'onlineSavingsMsg'
Learn More

TEMPO, TRC

Product Code. 30133267
Change view
Click to view available options
Quantity:
1 g
5 g
500 g
Unit Size:
1g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133267 5 g 5g
30135868 500 g 500g
30006302 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133267 Supplier Toronto Research Chemicals Supplier No. TRCT0178505G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2564-83-2
Molecular Formula C9 H18 N O
Molecular Weight (g/mol) 156.25
Synonym 1-Piperidinyloxy, 2,2,6,6-tetramethyl- (9CI, ACI), 2,2,6,6-Tetramethyl-1-piperidinyloxy (ACI), Piperidinooxy, 2,2,6,6-tetramethyl- (7CI, 8CI), 1,1,5,5-Tetramethylpentamethylene nitroxide, 1-Oxyl-2,2,6,6-tetramethylpiperidine, 2,2,6,6-Tetramethyl-1-oxylpiperidine, 2,2,6,6-Tetramethyl-1-piperadoxyl, 2,2,6,6-Tetramethyl-1-piperidine-N-oxyl, 2,2,6,6-Tetramethyl-1-piperidinoxyl, 2,2,6,6-Tetramethyl-1-piperidyloxy, 2,2,6,6-Tetramethylpiperidin-1-oxy, 2,2,6,6-Tetramethylpiperidin-1-oxyl radical, 2,2,6,6-Tetramethylpiperidin-N-oxyl, 2,2,6,6-Tetramethylpiperidine N-oxide, 2,2,6,6-Tetramethylpiperidine N-oxide radical, 2,2,6,6-Tetramethylpiperidine N-oxy, 2,2,6,6-Tetramethylpiperidine N-oxyl, 2,2,6,6-Tetramethylpiperidine N-oxyl radical, 2,2,6,6-Tetramethylpiperidine nitroxide, 2,2,6,6-Tetramethylpiperidine nitroxide radical, 2,2,6,6-Tetramethylpiperidine oxide, 2,2,6,6-Tetramethylpiperidine-1-oxyl, 2,2,6,6-Tetramethylpiperidino-1-oxy, 2,2,6,6-Tetramethylpiperidinooxy, 2,2,6,6-Tetramethylpiperidinooxy radical, 2,2,6,6-Tetramethylpiperidinooxyl, 2,2,6,6-Tetramethylpiperidinoxy, 2,2,6,6-Tetramethylpiperidinoxyl, 2,2,6,6-Tetramethylpiperidinoxyl radical, 2,2,6,6-Tetramethylpiperidinyl 1-oxide, 2,2,6,6-Tetramethylpiperidinyl-1-oxyl, 2,2,6,6-Tetramethylpiperidinyl-N-oxy, 2,2,6,6-Tetramethylpiperidinyloxy, 2,2,6,6-Tetramethylpiperidoxyl, HO 6, T 3751, Tanan, Tanane, Tempo, TMPO
IUPAC Name 1-λ^{1}-oxidanyl-2,2,6,6-tetramethylpiperidine
SMILES CC1(C)CCCC(C)(C)N1[O]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H18NO/c1-8(2)6-5-7-9(3,4)10(8)11/h5-7H2,1-4H3
Formula Weight 156.1388
Percent Purity >95%
Chemical Name or Material TEMPO
Grade HPLC
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.