missing translation for 'onlineSavingsMsg'
Learn More

TEMPO, free radical, 98+%

Product Code. 11440683
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440683 5 g 5g
11430683 1 g 1g
11450683 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440683 Supplier Thermo Scientific Alfa Aesar Supplier No. A12733.06
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2564-83-2 | C9H18NO | 156.25 g/mol

TEMPO is used as a catalyst in organic synthesis and for the oxidation of primary alcohols to aldehydes. It finds use in the chemical industry for conversion of bisnoralcohol (a steroid) to bisnoraldehyde. It acts as a free radical scavenger, as a mediator in controlled radical polymerization and as a structural probe in electron spin resonance spectroscopy. Further, it is involved in the preparation of (S)-(+)-2-methylbutanal from (S)-(-)-2-methyl-1-butanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TEMPO is used as a catalyst in organic synthesis and for the oxidation of primary alcohols to aldehydes. It finds use in the chemical industry for conversion of bisnoralcohol (a steroid) to bisnoraldehyde. It acts as a free radical scavenger, as a mediator in controlled radical polymerization and as a structural probe in electron spin resonance spectroscopy. Further, it is involved in the preparation of (S)-(+)-2-methylbutanal from (S)-(-)-2-methyl-1-butanol.

Solubility
Soluble in all organic solvents. Insoluble in water.

Notes
Store in a cool place. Incompatible withstrong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2564-83-2
Molecular Formula C9H18NO
Molecular Weight (g/mol) 156.25
MDL Number MFCD00009599
InChI Key QYTDEUPAUMOIOP-UHFFFAOYSA-N
Synonym 2,2,6,6-Tetramethyl-1-piperidinyloxy, free radical
IUPAC Name (2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
SMILES CC1(C)CCCC(C)(C)N1[O]

Specifications

Melting Point 31°C to 40°C
Boiling Point 193°C (decomposition)
Flash Point 67°C (152°F)
Quantity 5 g
UN Number UN3263
Beilstein 1422418
Merck Index 14,9140
Solubility Information Soluble in all organic solvents. Insoluble in water.
Formula Weight 156.25
Percent Purity ≥98%
Chemical Name or Material TEMPO, free radical
Show More Show Less
compliance-icons
Product Identifier
  • TEMPO, free radical
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2,2,6,6-Tetramethylpiperidinooxy

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.