missing translation for 'onlineSavingsMsg'
Learn More

Cytiva TempliPhi™ Sequence Resolver Kit

Uses modified nucleotides in isothermal rolling circle amplification (RCA) to improve results with difficult to sequence templates

Brand:  Cytiva 28-9035-29

Code : GX

Additional Details : Weight : 0.02500kg

 View more versions of this product

Product Code. 11339941

  • £377.00 / 1 set
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Includes: Sample buffer, Reaction buffer and Enzyme mix

Description

Description

  • Cultureless preparation from bacterial colonies, glycerol stock or purified DNA, saves time and reagents
  • Requires 20 minutes hands-on time
  • Excellent sequence finishing performance resolving sequencing stop and compression issues associated with difficult templates
  • Improve difficult template sequencing success with up to 820 bp Phred 20 read length
  • For use directly in cycle sequencing reactions without purification
  • Available in sizes of 20, 50 and 500 reactions

Specifications

Specifications

Store the kit at -70°C. The enzyme mix must be stored at -70°C; all other components may be stored at -20°CThaw components on ice and maintain at 0°C to 4°C during handling
Amplification of difficult templates for successful DNA sequencing
20
18 h
illustra TempliPhi Sequence Resolver Kit uses the highly processive, strand-displacing Phi29 DNA polymerase and modified nucleotides to amplify circular templates for subsequent sequencing.
1 x 90μL Sample Buffer, 1 x 90μL Reaction Buffer, 1 x 90μL Enzyme mix
illustra TempliPhi Sequence Resolver Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers