missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Temperature Restriction for UN75plus
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN75plus Oven
Unit Size:
Each
Description
- Accessory
- 1 Choice
- Temperatures: 60°C, 70°C, 80°C, 95°C, 100°C, 120°C, 160°C, 180°C, 200°C, or 250°C
Specifications
Specifications
| For Use With (Equipment) | UN75plus Oven |
| Type | Temperature Restriction |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction