Learn More
Memmert™ Temperature Profile Write/Read Unit (Pc/Chip Card) Up To 40 Ramps, For Ttc256
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
TTC256 Environmental Test Chamber
Unit Size:
Each
Description
- For programming via PC
- For writing to and reading from the chip card
- Up to 40 ramps
Specifications
Specifications
| Description | Up to 40 ramps, programming via PC |
| For Use With (Equipment) | TTC256 Environmental Test Chamber |
| Type | Temperature Profile Unit |