Learn More
OHAUS™ Temperature Probe
Hot plates and stirrers accessory
Brand:  OHAUS™ 30500593
Additional Details : Weight : 0.22700kg
 View more versions of this product
Product Code. 16620482
  • £303.41 / Each
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
Description
Description
  • Temperature probe in stainless steel or PTFE
  • Varying sizes available
Specifications
Specifications
25 cm
Temperature Probe
PTFE