Description
- Service includes 13 temperature measuring points and 1 humidity measuring point in center of walk-in chamber with a pair of values specified by the user
- Includes certificate
Specifications
Specifications
| For Use With (Equipment) | Binder™ Walk-in Chamber |
| Product Type | Measurement Service |