Learn More
BINDER™ Temperature and Humidity Calibration Certificate
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KBF PRO Series
KBF Series
Unit Size:
Each
Description
- 27 temperature measuring points
- 1 humidity measuring point, at 25°C/60% RH, or at specified values
Specifications
Specifications
| For Use With (Equipment) | KBF Series |
| Certifications/Compliance | DIN12880 |
| Duration | 1 Day |
| Product Type | Calibration Certificate |