Learn More
BINDER™ Temperature and Humidity Calibration Certificate
Product Code. 30238306 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KBF PRO Series
KBF Series
Unit Size:
Each
Product Code. 30238306
Brand: BINDER™ 80122578
Calibration certificate including 27 temperature measurements and 1 humidity measurement.
  • 27 temperature measuring points
  • 1 humidity measuring point, at 25°C/60% RH, or at specified values
Specifications
For Use With (Equipment) KBF PRO Series
Certifications/Compliance DIN12880
Duration 1 Day
Product Type Calibration Certificate