missing translation for 'onlineSavingsMsg'
Learn More

MAPA™ TempDex 710 Nitrile Glove

Product Code. p-7226653
Click to view available options
Size:
7
9
11
Unit Size:
10 pairs
This item is not returnable. View return policy

Product Code. 17668526

Brand: MAPA™ 34710037

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

TempDex 710 are for high dexterity and thermal protection.

  • Specific advantages
    • Thin, anatomically-shaped glove ensures dexterity
    • Suitable for long-term wear: excellent abrasion resistance
    • Silicone free to prevent traces, defects on metal sheet and glass prior to painting
    • Available in vending machine packaging
  • Specific features
    • Dot embossing for better insulation and better grip
  • Applications
    • Ceramic and plastics industry
      • Handling of composite materials after heat processing
    • Other industries
      • Handling of moulds in the ceramic industry
      • Handling of hot steel after welding
      • Handling of hot piping
    • Construction Industry (Heating engineers/Plumbers)
      • Maintaining pipes, heating equipment

Specifications

Applications Industrial
Color Yellow, Black
Material Nitrile
Size 7
For Use With (Application) Handling of Composite Materials After Heat Processing
Grip Design Dots
Lining Seamless Textile Support
Quantity 50/Cs.
Resistant To Abrasion
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.