missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Temozolomide

Product Code. 30117322 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30117322 5 g 5g
30117517 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30117322 Supplier Thermo Scientific Chemicals Supplier No. 466760050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85622-93-1 | C6H6N6O2, C6H6N6O2 | 194.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85622-93-1
Molecular Formula C6H6N6O2

Specifications

Quantity 5 g
Formula Weight 194.15
InChI Key BPEGJWRSRHCHSN-UHFFFAOYSA-N
SMILES CN1N=NC2=C(N=CN2C1=O)C(N)=O
IUPAC Name 3-methyl-4-oxo-3H,4H-imidazo[4,3-d][1,2,3,5]tetrazine-8-carboxamide
Molecular Weight (g/mol) 194.15
Chemical Name or Material Temozolomide
What is the recommended solubility of Temozolomide?

The solubility of temozolomide is 10 mg/mL in DMSO, which is what we recommend for reconstitution.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.