missing translation for 'onlineSavingsMsg'
Learn More

Temocapril hydrochloride, MedChemExpress

Product Code. p-200096381
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16484347

Brand: Medchem Express HYB0384

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Temocapril hydrochloride is an orally active angiotensin-converting enzyme (ACE) inhibitor. Temocapril hydrochloride can be used for the research of hypertension, congestive heart failure, acute myocardial infarction, insulin resistance, and renal diseases.

Temocapril hydrochloride is an orally active angiotensin-converting enzyme (ACE) inhibitor. Temocapril hydrochloride can be used for the research of hypertension, congestive heart failure, acute myocardial infarction, insulin resistance, and renal diseases.

Specifications

Chemical Name or Material Temocapril hydrochloride
Physical Form Powder
CAS 110221-44-8
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C23H29ClN2O5S2
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : 62.5 mg/mL (121.82 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 2.27 mg/mL (4.42 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(O)CN1C[C@@H](C2=CC=CS2)SC[C@H](N[C@H](C(OCC)=O)CCC3=CC=CC=C3)C1=O.Cl
Molecular Weight (g/mol) 513.07
Formula Weight 513.07
Percent Purity 99.63%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.