missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEMED, Ultrapure Bioreagent, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
50 mL
Packaging:
Amber Glass Bottle
Unit Size:
Each
Specifications
Specifications
| CAS | 110-18-9 |
| Flash Point | 10.00°C |
| Packaging | Amber Glass Bottle |
| Molecular Formula | C6H16N2 |
| Quantity | 50 mL |
| Specific Gravity | 1L = 0.78kg |
| Synonym | temed, n,n,n',n'-tetramethylethylenediamine, tmeda, 1,2-bis dimethylamino ethane, tetramethylethylenediamine, tetramethyldiaminoethane, tetrameen, propamine d, n1,n1,n2,n2-tetramethylethane-1,2-diamine, 1,2-ethanediamine, n,n,n',n'-tetramethyl |
| InChI Key | KWYHDKDOAIKMQN-UHFFFAOYSA-N |
| SMILES | CN(C)CCN(C)C |
| IUPAC Name | N,N,N',N'-tetramethylethane-1,2-diamine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction